Learn More
Potassium Cyanide, For Analysis, EMSURE™, MilliporeSigma™
Product Code. 15864490
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic Bottle
Unit Size:
250g
Product Code. Quantity Packaging Unit Size
15864490 250 g Plastic Bottle 250g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15864490 Supplier Supelco Supplier No. 1049670250
 Restricted Item
Forventet forsendelse: 19-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Specifications
Boiling Point 1625°C (1013hPa)
Melting Point 634°C
CAS 151-50-8
Color White
pH 11 to 12
Physical Form Solid
Quantity 250 g
Molecular Formula CKN
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
Solubility Information 716g/L
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
SMILES [C-]#N.[K+]
IUPAC Name potassium;cyanide
Molecular Weight (g/mol) 65.116
PubChem CID 9032
Formula Weight 65.12
Percent Purity >97.0%
Grade ACS
Packaging Plastic Bottle
Density 1.55 g/3 (20 °C)