Få mere at vide
Potassium Cyanide-13C,15N, TRC
Artikelnummer. 30408811
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30408811 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408811 Leverandør Toronto Research Chemicals Leverandørnr. TRCP695054100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Potassium Cyanide-13C,15N
CAS 74889-51-3
Molekylær formel 13C K 15N
Mængde 100 mg
Synonym Hydrocyanic-13C,15N acid','potassium salt, Potassium [13C,15N]cyanide, Potassium cyanide-13C,15N, Potassium cyanide-13C,15N
InChI formel InChI=1S/CN.K/c1-2;/i1+1,2+1;
SMIL [K][13C]#[15N]
IUPAC navn (^{15}N)azanylidyne(1^{13}C)methylpotassium
Molekylvægt (g/mol) 67.1
Formel vægt 66.9672