Få mere at vide
Potassium Chloride Solution 4M, Pure, Electrode Filling Solution, Fisher Chemical
Shop alle Fisher Chemical produkterBeskrivelse
100mL, Amber glass bottle
Tekniske data
Tekniske data
| CAS | 7447-40-7 |
| Fysisk form | Liquid |
| Mængde | 100 mL |
| Molekylær formel | ClK |
| MDL nummer | MFCD00011360 |
| Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
| InChI nøgle | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| SMIL | [Cl-].[K+] |
| IUPAC navn | potassium chloride |
| Molekylvægt (g/mol) | 74.55 |
| Vis mere |