Learn More
Potassium Carbonate, Supelco™
Product Code. 15864480
Change view
Click to view available options
Quantity:
1 kg
50 kg
Unit Size:
1kg
50kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15864480 1 kg 1kg
15837290 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15864480 Supplier Supelco Supplier No. 1049281000
 Restricted Item
Forventet forsendelse: 18-11-2026
For analysis EMSURE™ ACS,ISO,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 584-08-7
Specifications
Melting Point 891°C
Quantity 1 kg
Molecular Formula K2CO3
MDL Number MFCD00011382
Synonym Potassium carbonate, potash, Potassium carbonate
Molecular Weight (g/mol) 138.21
Grade Research
Density 2.43 g/cm3 at 20°C
Chemical Name or Material Potassium Carbonate