missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, ≥99.0%, Reagent Grade, Honeywell™

Catalog Number p-7120684
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
potassium;bromide
[K+].[Br-]
This item is not returnable. View return policy
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
potassium;bromide
[K+].[Br-]

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Boiling Point 1383°C (1013hPa)
Melting Point 734°C
Color Colorless
pH 5.0 to 8.0 (at 20°C)
Physical Form Solid
Quantity 500 g
Linear Formula KBr
UN Number NONH for all modes of transport
Formula Weight 119g/mol
Percent Purity ≥99.0%
Odor Odorless
Packaging Poly bottle
Vapor Pressure 1mmHg (795°C), 0.01mmHg (20°C)
Density 2.73g/cu.cm at 20°C
Chemical Name or Material Potassium bromide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'