missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium bis(trifluoromethylsulfonyl)imide

Brand:  Alfa Aesar™ 046909.03

Additional Details : CAS Number : 90076-67-8 Weight : 0.00100kg Transport : UN number : 1759 Chem class : 8 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 15472797

  • 745.05 DKK / 1g


In stock, ready to ship: 4
Add to basket

Specifications

Chemical Identifiers

90076-67-8
319.234
KVFIZLDWRFTUEM-UHFFFAOYSA-N
11099217
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[K+]
C2F6KNO4S2
MFCD06200829
potassium bistriflylimide anion, methanesulfonamide,1,1,1-trifluoro-n-trifluoromethyl sulfonyl-, potassium salt 1:1, potassium bis trifluoromethanesulfonly imide, ktfsi, potassium trifluoromethanesulfonimide, potassiobis trifluoromethylsulfonyl amine, potassium bis trifluoromethanesulfonyl amide, potassium bis trifluoromethyl sulfonyl imide, 1,1,1-trifluoro-n-potassio-n-trifluoromethanesulfonylmethanesulfonamide
potassium;bis(trifluoromethylsulfonyl)azanide

Specifications

Potassium bis (trifluoromethylsulfonyl)imide
198°C to 203°C
90076-67-8
(CF3SO2)2NK
UN1759
It is soluble in water. 10mg/mL.
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[K+]
319.234
319.24
Moisture sensitive
1g
C2F6KNO4S2
MFCD06200829
potassium bistriflylimide anion, methanesulfonamide,1,1,1-trifluoro-n-trifluoromethyl sulfonyl-, potassium salt 1:1, potassium bis trifluoromethanesulfonly imide, ktfsi, potassium trifluoromethanesulfonimide, potassiobis trifluoromethylsulfonyl amine, potassium bis trifluoromethanesulfonyl amide, potassium bis trifluoromethyl sulfonyl imide, 1,1,1-trifluoro-n-potassio-n-trifluoromethanesulfonylmethanesulfonamide
KVFIZLDWRFTUEM-UHFFFAOYSA-N
potassium;bis(trifluoromethylsulfonyl)azanide
11099217
Solid
Videos
SDS Documents