missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate, ≥99.0%, Reagent Grade, Honeywell™

Catalog Number 15638000
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
127-08-2
C2H3KO2
98.142
MFCD00012458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]
This item is not returnable. View return policy
127-08-2
C2H3KO2
98.142
MFCD00012458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point 292°C
Color Colorless
pH 7.5 to 8.5 (at 50.00g/L 20°C)
Physical Form Solid
Quantity 2.5 kg
Linear Formula CH3COOK
UN Number NONH for all modes of transport
Beilstein 3595449
Formula Weight 98.14g/mol
Percent Purity ≥99.0%
Odor Weak
Packaging Poly bottle
Density 1.57 g/cu.cm at 25°C
Chemical Name or Material Potassium acetate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'