missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acetate, ≥99.0%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | 292°C |
Color | Colorless |
pH | 7.5 to 8.5 (at 50.00g/L 20°C) |
Physical Form | Solid |
Quantity | 500 g |
Linear Formula | CH3COOK |
UN Number | NONH for all modes of transport |
Beilstein | 3595449 |
Formula Weight | 98.14g/mol |
Percent Purity | ≥99.0% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'