missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-n-butoxyphenyltrifluoroborate, 96%, Thermo Scientific™

Catalog Number p-7035375
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
850623-61-9
C10H13BF3KO
256.117
MFCD04115755
UEVIEKRZQJRPGE-UHFFFAOYSA-N
potassium 4-butoxyphenyl trifluoroborate, potassium 4-butoxyphenyltrifluorate, potassium 4-butoxyphenyl trifluoroboranuide, potassium 4-n-butoxyphenyl trifluorate, potassium 4-n-butoxyphenyl trifluoroborate, potassium 4-butoxyphenyl-trifluoroboranuide, potassium 4-butoxyphenyl trifluoro borate 1-, potassium 4-n-butoxyphenyltrifluoroborate, potassium ion 4-butoxyphenyl trifluoroboranuide, potassium 4-butoxyphenyl-tris fluoranyl boranuide
44717219
potassium;(4-butoxyphenyl)-trifluoroboranuide
[B-](C1=CC=C(C=C1)OCCCC)(F)(F)F.[K+]
This item is not returnable. View return policy
850623-61-9
C10H13BF3KO
256.117
MFCD04115755
UEVIEKRZQJRPGE-UHFFFAOYSA-N
potassium 4-butoxyphenyl trifluoroborate, potassium 4-butoxyphenyltrifluorate, potassium 4-butoxyphenyl trifluoroboranuide, potassium 4-n-butoxyphenyl trifluorate, potassium 4-n-butoxyphenyl trifluoroborate, potassium 4-butoxyphenyl-trifluoroboranuide, potassium 4-butoxyphenyl trifluoro borate 1-, potassium 4-n-butoxyphenyltrifluoroborate, potassium ion 4-butoxyphenyl trifluoroboranuide, potassium 4-butoxyphenyl-tris fluoranyl boranuide
44717219
potassium;(4-butoxyphenyl)-trifluoroboranuide
[B-](C1=CC=C(C=C1)OCCCC)(F)(F)F.[K+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 250 mg
Formula Weight 256.12
Percent Purity 96%
Chemical Name or Material Potassium 4-n-butoxyphenyltrifluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'