Få mere at vide
Potassium 3-chlorophenyltrifluoroborate, 96%, Thermo Scientific™
Artikelnummer. 15449907
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
Artikelnummer. 15449907
Brand: Thermo Scientific Alfa Aesar H53144.03
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 411206-75-2
Molekylær formel C6H4BClF3K
Molekylvægt (g/mol) 218.452
MDL nummer MFCD04115754
InChI nøgle LRJCVRYGKNDLAD-UHFFFAOYSA-N
Synonym potassium 3-chlorophenyl trifluoroborate, potassium 3-chlorophenyl trifluoroboranuide, potassium 3-chlorophenyltrifluoroborate, amtb707, potassium 3-chlorophenyl-trifluoroboranuide, potassium 3-chlorophenyl trifluoro borate 1-, potassium ion 3-chlorophenyl trifluoroboranuide, potassium 3-chlorophenyl-tris fluoranyl boranuide
PubChem CID 23677818
IUPAC navn potassium;(3-chlorophenyl)-trifluoroboranuide
SMIL [B-](C1=CC(=CC=C1)Cl)(F)(F)F.[K+]
Tekniske data
Kemisk navn eller materiale Potassium 3-chlorophenyltrifluoroborate
Smeltepunkt 76°C to 80°C
Mængde 1 g
Formel vægt 218.45
Procent renhed 96%