missing translation for 'onlineSavingsMsg'
Learn More

Potassium 3,4-dichlorophenyltrifluoroborate, 96%, Thermo Scientific™

Catalog Number p-7035502
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
850623-68-6
C6H3BCl2F3K
252.894
MFCD04115763
RTZOKQYJVLRZFW-UHFFFAOYSA-N
potassium 3,4-dichlorophenyl trifluoroborate, potassium 3,4-dichlorophenyl trifluoroboranuide, potassium 3,4-dichlorophenyltrifluoroborate, c6h3bcl2f3.k, potassium 3,4-dichlorophenyl-trifluoroboranuide, potassium ion 3,4-dichlorophenyl trifluoroboranuide, potassium 3,4-dichlorophenyl trifluoro borate 1-, potassium 3,4-dichlorophenyl-tris fluoranyl boranuide
44717229
potassium;(3,4-dichlorophenyl)-trifluoroboranuide
[B-](C1=CC(=C(C=C1)Cl)Cl)(F)(F)F.[K+]
This item is not returnable. View return policy
850623-68-6
C6H3BCl2F3K
252.894
MFCD04115763
RTZOKQYJVLRZFW-UHFFFAOYSA-N
potassium 3,4-dichlorophenyl trifluoroborate, potassium 3,4-dichlorophenyl trifluoroboranuide, potassium 3,4-dichlorophenyltrifluoroborate, c6h3bcl2f3.k, potassium 3,4-dichlorophenyl-trifluoroboranuide, potassium ion 3,4-dichlorophenyl trifluoroboranuide, potassium 3,4-dichlorophenyl trifluoro borate 1-, potassium 3,4-dichlorophenyl-tris fluoranyl boranuide
44717229
potassium;(3,4-dichlorophenyl)-trifluoroboranuide
[B-](C1=CC(=C(C=C1)Cl)Cl)(F)(F)F.[K+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 178°C to 180°C
Quantity 1 g
Formula Weight 252.9
Percent Purity 96%
Chemical Name or Material Potassium 3,4-dichlorophenyltrifluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'