missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2-methoxyphenyltrifluoroborate, 96%, Thermo Scientific™

Catalog Number p-7034882
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
236388-46-8
C7H7BF3KO
214.04
MFCD04115756
IXJPOOXJHKETAZ-UHFFFAOYSA-N
potassium 2-methoxyphenyl trifluoroborate, potassium trifluoro 2-methoxyphenyl borate, potassium 2-methoxyphenyltrifluoroborate, potassium trifluoro 2-methoxyphenyl boranuide, pubchem11466, amtb101, trifluoro 2-methoxyphenyl potassioboron v, potassium trifluoro-2-methoxyphenyl boranuide, potassium trifluoro 2-methoxyphenyl borate 1-
23684966
potassium;trifluoro-(2-methoxyphenyl)boranuide
[K+].COC1=CC=CC=C1[B-](F)(F)F
This item is not returnable. View return policy
236388-46-8
C7H7BF3KO
214.04
MFCD04115756
IXJPOOXJHKETAZ-UHFFFAOYSA-N
potassium 2-methoxyphenyl trifluoroborate, potassium trifluoro 2-methoxyphenyl borate, potassium 2-methoxyphenyltrifluoroborate, potassium trifluoro 2-methoxyphenyl boranuide, pubchem11466, amtb101, trifluoro 2-methoxyphenyl potassioboron v, potassium trifluoro-2-methoxyphenyl boranuide, potassium trifluoro 2-methoxyphenyl borate 1-
23684966
potassium;trifluoro-(2-methoxyphenyl)boranuide
[K+].COC1=CC=CC=C1[B-](F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 282°C to 286°C
Quantity 1 g
Formula Weight 214.04
Percent Purity 96%
Chemical Name or Material Potassium 2-methoxyphenyltrifluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'