Få mere at vide
Potassium 2-hydroxy-4-methoxybenzoate, TRC
Artikelnummer. 30411211
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30411211 500 mg
30421761 100 mg
30423538 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30411211 Leverandør Toronto Research Chemicals Leverandørnr. TRCP994268500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 152312-71-5
Molekylær formel C8H7KO4
Molekylvægt (g/mol) 206.24
IUPAC navn potassium;2-hydroxy-4-methoxybenzoate
SMIL [K+].COc1ccc(C(=O)[O-])c(O)c1
Tekniske data
Kemisk navn eller materiale Potassium 2-hydroxy-4-methoxybenzoate
Mængde 500 mg
InChI formel InChI=1S/C8H8O4.K/c1-12-5-2-3-6(8(10)11)7(9)4-5;/h2-4,9H,1H3,(H,10,11);/q;+1/p-1
Formel vægt 205.9981