Learn More
Gilson™ Position Rack Tube Vessel Type, Fraction Collector
These racks are for use with the FC203B and FC204 Fraction Collectors
269.24 DKK - 6868.80 DKK
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | ||||
16676105
|
Gilson™
12040101 |
9 mL Vial (13 x 100 mm) | Code 1 |
269.24 DKK
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
16626115
|
Gilson™
130412 |
5 mL Vial (12 x 75 mm) | Code 4 |
6868.80 DKK
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
Description
- HPLC Purification Technique
- Maximum Vessel Quantity is 80
- Tube Vessel Type
Specifications
80 |