missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Porcine Liver Esterase, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Porcine Liver Esterase |
| CAS | 9016-18-6 |
| Mængde | 1 g |
| Synonym | Carboxylesterase, 1,4-Butanediol Diacrylate Esterase, 7-Amino-3-methoxy-3-cephem-4-carboxyl Ester Hydrolase, Aliesterase, Aminoacyl Esterase, B-Esterase, Biocatalytics 123, Butyrate Esterase, Butyryl Esterase, C4 Esterase, Carbonic Esterase, Carboxyesterase, Carboxyl Ester Hydrolase, Carboxyl Ester Lipase, Carboxyl Esterase, Carboxylate Esterase, Carboxylesterase 1, Carboxylesterase 2, Carboxylesterase B, Carboxylesterase ES-1, Carboxylesterases, Carboxylic Acid Esterase, Carboxylic Ester Hydrolase, Carboxylic Esterase, Chirazyme E 1, Chirazyme E-2, Chirazyme E-3, Cinnamate Esterase, Cinnamic Acid Esterase, Cinnamoyl Ester Hydrolase, Cinnamoyl Esterase, E 0231, E.C. 3.1.1.1, E.C. 3.1.1.12, EC 3.1.1.1, Egasyn, Esterase (EC 3.1.1.1), Esterase 29, Esterase 6, Esterase C4, Esterase D (E.C. 3.1.1.1), Esterase D (EC 3.1.1.1), Esterase EP10, Esterase, B-, Esterase-30, Fatty Acid Ethyl Ester Hydrolase, Fluazifop-butyl Esterase, Ketoprofen Alkyl Esterase, Ketoprofen Choline Esterase, Methyl Farnesoate Esterase, Methylbutyrase, Methylbutyrate Esterase, Monobutyrase, Naproxen Esterase, Neutral Esterase, Nonspecific Carboxylesterase, Paraben Esterase, Phthalate Ester Hydrolase, Phthalate Esterase, Pig Liver Esterase, Procaine Esterase, Propionyl Esterase, Prostaglandin A2 Methylesterase, Egasyns Proteins, SCA 50, SNSM 87, Short Chain Fatty Acid Esterase, Sterase, TAME Esterase, Texazyme PES Sp5, Thiazopyr Esterase, Triacetin Esterase, Valacyclovirase, Vitamin A Esterase, α-Carboxylesterase, α-Esterase, β-Carboxylesterase, β-Esterase, δ-Aminolevulinate Esterase |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?