Learn More
Karl Hecht™ Porcelain Half-Round Evaporating Dishes
Glazed inside and outside for an exceptionally smooth finish.
107.06 DKK - 274.54 DKK
Specifications
Description | Half-round |
---|---|
Bottom Shape | Flat |
Material | Porcelain |
Shape | Round |
Type | Evaporating Dish |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10195631
|
Karl Hecht™
47000007 |
50 mL |
107.06 DKK
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
10359271
|
Karl Hecht™
47000008 |
75 mL |
122.96 DKK
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
10033971
|
Karl Hecht™
47000012 |
310 mL |
274.54 DKK
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Description
- With spout
- Porcelain construction
- DIN 12903
- Available in a range of capacities
Specifications
Half-round | |
Porcelain | |
Evaporating Dish |
Flat | |
Round | |
Glazed |