missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POP4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12413
Additional Details : Weight : 0.00970kg
Description
POP4 Polyclonal antibody specifically detects POP4 in Human, Porcine, Bovine, Feline, Primate samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
POP4 | |
Polyclonal | |
Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide | |
Rabbit | |
Affinity-purified | |
RUO | |
Primary | |
Human, Porcine, Bovine, Feline, Primate | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
EC 3.1.26.5, processing of precursor 4, ribonuclease P/MRP subunit (S. cerevisiae), ribonuclease P protein subunit p29, RPP29hPOP4 | |
Synthetic peptide taken within amino acid region 140-190 on human Ribonuclease P protein subunit p29 protein. | |
100 μg | |
Endocrinology | |
10775 | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |