missing translation for 'onlineSavingsMsg'
Learn More

Ponceau S, Electrophoresis Grade, Thermo Scientific Chemicals

Catalog Number p-7041925
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
6226-79-5
C22H12N4Na4O13S4
760.552
MFCD00003892
VSXKEUCERCWGKF-STNZDNLRSA-J
C.I. 27195
11320219
tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
This item is not returnable. View return policy
6226-79-5
C22H12N4Na4O13S4
760.552
MFCD00003892
VSXKEUCERCWGKF-STNZDNLRSA-J
C.I. 27195
11320219
tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Protein stain for electrophoresis

Ponceau S dye is used to make a stain for rapid reversible staining of protein bands on nitrocellulose or PVDF membranes (Western blots) and also for staining protein on cellulose acetate membranes. It is also suitable for electrophoresis, azo dye for reversible protein staining. This stain can also be used as a fiducial marker in mass spectrometric imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ponceau S dye is used to make a stain for rapid reversible staining of protein bands on nitrocellulose or PVDF membranes (Western blots) and also for staining protein on cellulose acetate membranes. It is also suitable for electrophoresis, azo dye for reversible protein staining. This stain can also be used as a fiducial marker in mass spectrometric imaging.

Solubility
Soluble in water (10 g/L) at 20°C. Insoluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Solubility Information Soluble in water (10g/L) at 20°C. Insoluble in ether.
Formula Weight 760.58
Grade Electrophoresis
Color Brown to Red
Quantity 10 g
Physical Form Powder
Chemical Name or Material Ponceau S

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'