Få mere at vide
MedChemExpress Pomalidomide-PEG4-C2-NH2
Artikelnummer. 16471606
Encompass_Preferred
Skift visning
Mængde:
100 mg
500 mg
1 g
2 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16471606 500 mg
16451179 1 g
16420146 2 g
16405496 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16471606 Leverandør MedChemExpress Leverandørnr. HY112599
MedChemExpress Pomalidomide-PEG4-C2-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and 4-unit PEG linker used in PROTAC technology.
Tekniske data
Molekylvægt (g/mol) 492.52 g/mol
CAS 2225940-52-1
Indhold og opbevaring Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Forsendelsestilstand Room temperature
Mængde 500 mg
Synonym Cereblon Ligand-Linker Conjugates 8 ∣ E3 Ligase Ligand-Linker Conjugates 22
Opløselighed DMSO : 50 mg/mL (101.52 mM; Need ultrasonic)