Få mere at vide
Pomalidomide-(d4 major), TRC
Artikelnummer. 30414299
Skift visning
Mængde:
0.25 mg
Artikelnummer. Mængde
30414299 0.25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414299 Leverandør Toronto Research Chemicals Leverandørnr. TRCP6882020.25MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pomalidomide-d4
CAS 1416575-78-4
Molekylær formel C13H7D4N3O4
Mængde 0.25 mg
Synonym 4-Amino-2-(2,6-dioxo-3-piperidinyl)isoindole-1,3-dione-d4, 4-Amino-2-(2,6-dioxo-3-piperidyl)isoindoline-1,3-dione-d4, Actimid-d4, CC 4047-d4, IMiD 3-d4
InChI formel InChI=1S/C13H11N3O4/c14-7-3-1-2-6-10(7)13(20)16(12(6)19)8-4-5-9(17)15-11(8)18/h1-3,8H,4-5,14H2,(H,15,17,18)/i4D2,5D2
SMIL O=C(C(C([2H])([2H])C1([2H])[2H])N2C(C3=CC=CC(N)=C3C2=O)=O)NC1=O
Molekylvægt (g/mol) 277.27
Formel vægt 277.1001