Få mere at vide
Tocris Bioscience™ Pomalidomide 4'-PEG5-amine
Beskrivelse
Pomalidomide 4'-PEG5-amine is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG5 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-[(17-Amino-3,6,9,12,15-pentaoxaheptadec-1-yl)amino]-2-(2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione hydrochloride |
| CAS | 2357117-23-6 |
| Mængde | 25 mg |
| Mål | Degrader Building Blocks |
| Molekylær formel | C25H36N4O9.HCl |
| Renhed | 95% |