Få mere at vide
Tocris Bioscience™ Pomalidomide 4'-alkylC6-azide
Beskrivelse
Pomalidomide 4'-alkylC6-azide is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC6 linker with terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-[(6-Azidohexyl)amino]-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione |
| CAS | 2375555-72-7 |
| Mængde | 25 mg |
| Mål | Degrader Building Blocks |
| Molekylær formel | C19H22N6O4 |
| Renhed | 0.95 |