Få mere at vide
Tocris Bioscience™ Pomalidomide 4'-alkylC5-amine
Beskrivelse
Pomalidomide 4'-alkylC5-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC5 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was Pomalidomide - linker 4
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-[(5-Aminopentyl)amino]-2-(2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione hydrochloride |
| CAS | 2375194-03-7 |
| Synonym | Pomalidomide - linker 4 |
| Mængde | 25 mg |
| Molekylær formel | C18H22N4O4.HCl |
| Formel vægt | 394.86 |
| Produkttype | Ligand |
| Renhed | 95% |