Få mere at vide
Kartell™ POM Slide Staining Trough
Beskrivelse
Stain up to 25 slides with a versatile trough that can also be used as a mailing box. Kartell™ POM Slide Staining Trough is constructed of POM and designed to hold up to 25 slides.
- Holds up to 25 slides
- Accommodates one Kartell™ 1100 rack
- Can also be used as a mailing box
Tekniske data
Tekniske data
| Produkttype | Staining Trough |
| Slidekapacitet | 25 |
| Til brug med (udstyr) | Slides |
| Dimensioner (L x B x H) | 88 x 100.8 x 52.4 mm |
| Højde (metrisk) | 52,4 mm |