Learn More
Macherey-Nagel™ Chromafil™ Polyvinylidene Difluoride Syringe Filters
For aqueous samples, water-soluble oligomers and polymers like proteins. Macherey Nagel™ Chromafil™ Polyvinylidene Difluoride Syringe Filters are labeled for method validation and certification.
Brand: Macherey-Nagel™ 729039
Additional Details : Weight : 0.01000kg
Description
- Hydrophilic membrane
- For aqueous samples, water-soluble oligomers and polymers like proteins
- Binding capacity for proteins 20 μg per 25 mm filter
Specifications
Top Black, Bottom White | |
1.0/0.45 μm | |
Polypropylene (PP) | |
100 Pack | |
∼80 μL |
PVDF with Glass Fiber Prefilter | |
∼4.9 cm2 | |
Glass Fibre | |
Syringe Filter | |
25 mm |