missing translation for 'onlineSavingsMsg'
Learn More

Polyvinyl alcohol, 87-89% hydrolyzed, high molecular weight, Thermo Scientific Chemicals

Catalog Number p-7051452
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
9002-89-5
(C2H4O)n
44.05
MFCD00081922
IMROMDMJAWUWLK-UHFFFAOYSA-N
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
11199
ethenol
OC(-*)C-*
This item is not returnable. View return policy
9002-89-5
(C2H4O)n
44.05
MFCD00081922
IMROMDMJAWUWLK-UHFFFAOYSA-N
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
11199
ethenol
OC(-*)C-*

Polyvinyl alcohol is used as a biological reagent to test the dispersion of zinc oxide nanoparticles. It is an effective protein resistant material and is used to stabilize gold colloids of diameter 3-8nm. It is used as an emulsion polymerization aid to make polyvinyl acetate, as a protective colloid, as a film used in the water transfer printing process, as a surfactant for the formation of polymer encapsulated nanobeads, as an embolization agent in medical procedures and as a fixative for specimen collection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Solid
Solubility Information Soluble in water.
Quantity 25 g
Percent Purity 87 to 89%
Odor Odorless
Chemical Name or Material Polyvinyl alcohol, High molecular weight, Hydrolyzed

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'