Få mere at vide
Polythiazide, LoGiCal
Artikelnummer. 30460471
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460471 10 mg
1 options
Artikelnummer. 30460471 Leverandør LoGiCal Leverandørnr. LGCFOR3231.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Polythiazide
CAS 346-18-9
Analyt- eller komponentnavne Polythiazide
Forsendelsestilstand Room Temperature
Molekylær formel C11 H13 Cl F3 N3 O4 S3
Certificeringer/compliance ISO 9001
Synonym 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 6-chloro-3,4-dihydro-2-methyl-3-[[(2,2,2-trifluoroethyl)thio]methyl]-, 1,1-dioxide, (±)-Nephril, (±)-Polythiazide, 2-Methyl-3-(β,β,β-trifluoroethylthiomethyl)-6-chloro-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine-1,1-dioxide, 6-Chloro-3,4-dihydro-2-methyl-3-[[2,2,2-trifluoroethyl)thio]methyl]-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 6-Chloro-3,4-dihydro-2-methyl-7-sulfamoyl-3-(2,2,2-trifluoroethylthiomethyl)-2H-1,2,4-benzothiadiazine 1,1-dioxide, Drenusil, NSC 108161, Nephril, P 2525, Polythiazide, Renese
InChI formel InChI=1S/C11H13ClF3N3O4S3/c1-18-10(4-23-5-11(13,14)15)17-7-2-6(12)8(24(16,19)20)3-9(7)25(18,21)22/h2-3,10,17H,4-5H2,1H3,(H2,16,19,20)
Løsningstype Neat
SMIL CN1C(CSCC(F)(F)F)Nc2cc(Cl)c(cc2S1(=O)=O)S(=O)(=O)N
IUPAC navn 6-chloro-2-methyl-1,1-dioxo-3-(2,2,2-trifluoroethylsulfanylmethyl)-3,4-dihydro-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 439.88
Mængde 10 mg
Formel vægt 438.9709 g/mol