Få mere at vide
Globe Scientific Polystyren multifunktionelle reagensglas
Artikelnummer. 31922539 Shop alle Globe Scientific produkter
Skift visning
Emballage:
1000/Bag, 1 Bag/Box
250/Bag, 8 Bags/Case
250/Oriented Box, 1000/Cs.
Bag of 1000
Farve:
Clear
Light Blue
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Emballage Farve
31922539 1000/Bag, 1 Bag/Box Clear
31922607 250/Oriented Box, 1000/Cs. Clear
31923468 250/Bag, 8 Bags/Case Clear
31922492 Bag of 1000 Light Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 31922539 Leverandør Globe Scientific Leverandørnr. 110416
Forventet forsendelse: 11-11-2026
Formet af polystyren i premium-kvalitet for klarhed
  • Multi-use tubes
  • Can withstand 1500rpm
Tekniske data
Farve Clear
Kapacitet (metrisk) 5 mL
Længde (metrisk) 75 mm
Materiale Polystyrene
Emballage 1000/Bag, 1 Bag/Box
Diameter (metrisk) 12 mm
Produkttype Test Tube