Learn More
Polystyrene latex microsphere, 0.75 micron, 2.5 wt% dispersion in water
Product Code. 15476515
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
Product Code. Quantity Unit Size
15476515 15 mL 15mL
1 options
Product Code. 15476515 Supplier Thermo Scientific Alfa Aesar Supplier No. 042742.4Y
 Restricted Item
På lager
Add to basket
CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Polystyrene latex microsphere is used as a biocompatible material, especially in dentistry. They are used as insulators, for injection molding and casting, as sheets, plates, rods, rigid forms and beads. It is also used as extrusion molded foamed packaging, dinnerware, bottles, toys and disposable cutlery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 9003-53-6
Physical Form Microsphere
Quantity 15 mL
Odor Odorless
Molecular Formula (C8H8)n
Particle Size 0.75 Micron
MDL Number MFCD00084450
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
Solubility Information Fully miscible with water.
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
SMILES *-CC(-*)C1=CC=CC=C1
IUPAC Name ethenylbenzene
Molecular Weight (g/mol) 104.15
PubChem CID 7501
ChEBI CHEBI:27452
Concentration or Composition (by Analyte or Components) 2.5 wt% dispersion in water
Chemical Name or Material Polystyrene latex microsphere