Learn More
Technical Service Consultants™ Polystyrene L-shaped Spreader
Product Code. 12739885
Change view
Click to view available options
Quantity:
1000/Pk.
500 Pk.
Unit Size:
Pack of 1000
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12739885 1000/Pk. Pack of 1000
12749885 500 Pk. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12739885 Supplier Technical Service Consultants™ Supplier No. TS/30A
  • ONLINE EXCLUSIVE PRICE
    1500.00 DKK
    1263.67 DKK / Pakke med 1000
    Spare 236.33 DKK
    15% Slukket
Forventet forsendelse: 22-12-2026
Use Technical Service Consultants™ Polystyrene L-shaped Spreader for surface spreading of liquid or culture samples uniformly over the surface of solid media. Shaped like a hockey stick, it is recommended for single use practices or sterile techniques.
  • Polystyrene
  • Three year shelf life
  • Ideal for bacterial counts
  • L shaped plastic (firm) peel pouch
  • Hockey Stick Shape
Specifications
Quantity 1000/Pk.
Shape L-Shape
Material Polystyrene
Product Type Spreader
Sterility Sterile