Få mere at vide
Sampling Systems™ SteriWare™ Engangs PowderSafe
Shop alle Sampling Systems produkter
Sterilitet:
Ikke steril
Steril
Pakningsstørrelse:
Pakke med 100
Beskrivelse
Key Features:
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Supplied with see-through container that securely clips over the spatula
Why Use a SteriWare™ Disposable Sampler?
- No Cross Contamination - All SteriWare™ devices can be used straight from the bag.
- No Cleaning - Unlike a reusable device, you can use a SteriWare™ product straight away. This eliminates the need for costly Cleaning Validation.
- High Quality - Our SteriWare™ products are manufactured/moulded/packed in a Class 100,000 Cleanroom in England. Please see the products Certificate of Conformity and Technical Specification Sheet for more information.
- Full Documentation - We can provide full batch specific documentation for all SteriWare™ products. This includes Certificates of Conformity and Certificates of Irradiation (where applicable). The batch number is clearly printed on every bag and box.
- Cost Effective - Removing the need for Cleaning Validation by switching to a SteriWare™ device saves time and money.
- Available Sterile - Most SteriWare™ products are available pre-sterilized by Gamma or Electron Beam to SAL10-6. Please see the Specification Sheet for more information.
Tekniske data
Tekniske data
| Sterilitet | Steril |
| Farve | White Spatula, Transparent Cover |
| Certificeringer/compliance | Conforms to FDA CFR 177.1640, Conforms to EU Regulation 10/2011 |
| Engangs | Single-use |
| Produkttype | Powder Sampler |
| Til brug med (applikation) | For Easy, Controlled Powder Discharge |
| Materiale | Polystyrene |
| Længde (metrisk) | 230 mm |