missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Polyquaternium-37, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Polyquaternium-37 |
| CAS | 26161-33-1 |
| Molekylær formel | (C9H18ClNO2)x |
| Mængde | 500 mg |
| Synonym | Choline Chloride Methacrylate PolymersN,N,N-Trimethyl-2-[(2-methyl-1-oxo-2-propenyl)oxy]ethanaminium Chloride HomopolymerMethacrylic Acid Ester With Choline Chloride Polymers, (Methacryloxyethyl)trimethylammonium Chloride Polymer, 2-(Methacryloyloxy)ethyl Trimethylammonium Chloride Homopolymer, 2-(Trimethylammonium)ethyl Methacrylate Chloride Homopolymer, Alcostat 567, Cosmedia CTH(E), Dimethylaminoethyl Methacrylate Methyl Chloride Homopolymer, Dimethylaminoethyl Methacrylate-methyl Chloride Copolymer, Evagrowth C 104G, Flocogil C 4, Himoloc MP 173H, Himoloc NR 70, Himoloc NR 703, K 415, Kayafloc C 599, Light Ester DQ 100 Homopolymer, Methacroylcholine Chloride Homopolymer, PMETAC, Poly(2-methacryloxyethyltrimethylammonium Chloride), Poly(2-methacryloyloxyethyltrimethylammonium Chloride), Poly(2-trimethylammonioethyl Methacrylate Chloride), Poly(N,N,N-trimethylaminoethyl Methacrylate Chloride), Poly(dimethylaminoethyl Methacrylate Methyl Chloride), Poly(methacryloxyethyltrimethylammonium Chloride), Poly(methacryloyloxyethyltrimethylammonium Chloride), Poly(methylacryloyloxyethyltrimethyl Sodium Chloride), Poly(β-methacryloxyethyltrimethylammonium Chloride), PolyTMMC, Poly[(trimethylammonio)ethyl Methacrylate Chloride], Poly[N,N,N-trimethyl-N-(2-methacryloxyethyl)ammonium Chloride], Poly[[β-(methacryloyloxy)ethyl]trimethylammonium Chloride], Poly[trimethyl(β-methacryloyloxyethyl)ammonium Chloride], Poly[trimethyl[2-(methacryloyloxy)ethyl]ammonium Chloride], Praestol 444K, QDM WFixer 013, Rheocare CTH(E), Rheocare Ultragel 300, Salcare SC 95, Sanfloc C 009P, Shallol, Synthalen CR, Takiflock C 409, Trimethylaminoethyl Methacrylate Chloride Homopolymer, Trimethylammonioethyl Methacrylate Chloride Homopolymer, Trimethylammonioethyl Methacrylate Chloride Polymer, Ultragel 300, Zetag 88N, [2-(Methacroyloxy)ethyl]trimethylammonium Chloride Homopolymer, β-Methacryloyloxyethyltrimethylammonium Chloride Polymer |
| InChI formel | InChI=1S/C9H18NO2.ClH/c1-8(2)9(11)12-7-6-10(3,4)5;/h1,6-7H2,2-5H3;1H/q+1;/p-1 |
| SMIL | O=C(C(C)=C)OCC[N+](C)(C)C.[Cl-] |
| IUPAC navn | 2-(methacryloyloxy)-N,N,N-trimethylethan-1-aminiumchlorid |
| Molekylvægt (g/mol) | 207.7 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?