Learn More
BRAND™ Polypropylene Valve Cap
For closing valve block. Suitable for autoclaving.
218.00 DKK
Specifications
Product Type | Valve Cap |
---|---|
Autoclavable | Autoclavable |
Material | Polypropylene |
For Use With (Equipment) | Seripettor™ / Seripettor™ pro |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10673392
|
BRAND™
704554 |
25 mL |
218.00 DKK
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
10039800
|
BRAND™
704552 |
2, 10 mL |
218.00 DKK
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
- Polypropylene construction
- Autoclavable
- For use with seripettor™
Specifications
Valve Cap | |
Autoclavable | |
Polypropylene | |
Seripettor™ / Seripettor™ pro |