Learn More
Cole-Parmer™ Polypropylene Disposable Spatulas, Opaque
Product Code. 11755219 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (Metric):
140 mm
222 mm
Unit Size:
Pack of 300
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11755219 140 mm Pack of 300
11989698 222 mm Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11755219 Supplier Cole-Parmer™ Supplier No. 0626514
  • ONLINE EXCLUSIVE PRICE
    497.00 DKK
    464.61 DKK / Pakke med 300
    Spare 32.39 DKK
    6% Slukket
Forventet forsendelse: 10-12-2026
Reduce cross-contamination with these polypropylene disposable spatulas. Polypropylene Disposable Spatulas are ideal for chemical and biological material transfer, collection, and manipulation. Spatulas feature both a scoop end and a spatula end.
  • Disposable polypropylene (PP) spatulas
  • Use as a spatula, stir rod, or policeman
  • Contoured end is shatter resistant even when exposed to liquid nitrogen
  • Flexible enough to fit into many small containers to scrape or collect samples
  • Hollow core reduces heat and cold transfer
  • Can be used with dilute acids and bases
  • Autoclavable
Specifications
Quantity 300 Pk.
Autoclavable Autoclavable
Shape Scoop End, Spatula End
Color Opaque
Disposable Single-use
Product Type Spatula
For Use With (Application) For chemical and biological material transfer, collection, and manipulation, used with dilute acids and bases, reduce cross-contamination
Material Polypropylene
Length (Metric) 140 mm