Learn More
Cytiva Polypropylene On-line Filter Kit
Used for AKTA chromatography systems.  Cytiva Polypropylene On-line Filter Kits are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules.
Brand:  Cytiva 18-1027-11
846.45 DKK valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Additional Details : Weight : 0.01080kg
Product Code. 10720740
  • 891.00 DKK / Pack of 10
Estimated Shipment: 03-05-2024
to see stock.
Add to basket
Description
Description
  • Kit contains 10 x filter 2 x Support screen
  • The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran
  • Polypropylene (PP) 
Specifications
Specifications
4°C
10 Pack