Learn More
BRAND™ Polypropylene Micro Tubes Cap Insert
Product Code. 10480243 Shop All Brand Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Pack of 500
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10480243 Blue Pack of 500
10548931 Red Pack of 500
10324851 Yellow Pack of 500
10028960 Green Pack of 500
15527851 White Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10480243 Supplier BRAND™ Supplier No. 780721
  • ONLINE EXCLUSIVE PRICE
    158.00 DKK
    149.26 DKK / Pakke med 500
    Spare 8.74 DKK
    5% Slukket
På lager
Polypropylene Micro Tubes Cap Insert for use with all non-tamper evident screw caps allows for easy identification.
  • For micro tubes with attached or bulk screw caps
  • Suitable for use with non-tamper evident caps
TRUSTED_SUSTAINABILITY
Specifications
Product Type Microtube Cap Insert
Material Polypropylene
Color Blue
Temperature Range (Metric) -196°C to +121°C
For Use With (Equipment) Microtubes, Screw Caps