Få mere at vide
Polypropylene Lift Pumps
Artikelnummer. 12917974
Encompass
Oversigt:
Includes 1000 mm Downtube
Includes 900 mm Downtube
Pakningsstørrelse:
Hver
Artikelnummer. 12917974
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
For 20L/25L containers, capacity 0.33L per stroke. For 205L containers capacity 0.5L per stroke. Available in two sizes. With a 900mm flexible dispensing tube, except polypropylene 205L dispensing pump.
  • Manufactured with nylon for maximum resistance to acids and solvents
  • No metal part comes into contact with the liquid being pumped
  • PTFE seals fitted
  • Universal adapters fitted to provide secure seating in the necks of virtually all drums from 20L/25L to 205L capacities
  • Piston action ensures that the liquid flow is totally under control at all times during the action For dispensing solvents and acids in controlled amounts.
Tekniske data
Oversigt Includes 900 mm Downtube
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.