Få mere at vide
Masterflex™ Polypropylene Cross Connectors
Beskrivelse
- Polypropylene cross connectors, 10mm
- Barbed fittings are designed for quick and easy assembly without tools
- Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
- Available in packs of 20
Tekniske data
Tekniske data
| Autoklaverbar | Autoclavable |
| Materiale | Polypropylene |
| Diameter (metrisk) Udvendig | 10 mm |
| Samlet længde (metrisk) | 50 mm |
| Emballagemængde | 20 Fittings |
| Slangestørrelse | 10mm ID. |
| Steriliseringsmetode | Autoclave |
| Form | Cross |
| Farve | White |
| Produkttype | Cross Connectors |