Learn More
VITLAB™ Polypropylene Bulb Pipet
Brand:  VITLAB™ 164494
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10031462
  • 1296.28 DKK / Pack of 6
Estimated Shipment: 04-01-2023
to see stock.
Specifications
Yes
6 Pack
450 mm
Polypropylene
25 mL
Bulb Pipette