Learn More
Sampling Systems™ Polypropylene Braided Cord
Sampling Systems™ Polypropylene Braided Cord is used to help lower the bucket of the Enviro Dipper.
148.00 DKK - 655.00 DKK
Specifications
Product Type | Braided Cord |
---|---|
Material | Polypropylene |
For Use With (Equipment) | Enviro Dipper |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
15839351
|
Sampling Systems™
5070P-10 |
10 m |
148.00 DKK
10m |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
15849351
|
Sampling Systems™
5070P-25 |
25 m |
340.00 DKK
25m |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
15859351
|
Sampling Systems™
5070P-50 |
50 m |
655.00 DKK
50m |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
Description
- For use with an Enviro Dipper
- Polypropylene construction
Specifications
Braided Cord | |
Polypropylene | |
Enviro Dipper |