Learn More
Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Thread
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Thread enables smaller containers to be supported on top the existing reservior.
Brand:  Poulten & Graf GmbH 101 09025
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10610713
  • 88.40 DKK / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • Suitable for OPTIFIX and POLYFIX dispenser
  • Made of Polypropylene
  • Adapter with threads
  • Can be used for most chemicals and liquids
Specifications
Specifications
Adapters for OPTIFIX and POLYFIX Dispensers
White
Polypropylene
1
Autoclavable
Adapters for OPTIFIX and POLYFIX dispensers
OPTIFIX and POLYFIX dispensers
GL 25