Learn More
Cole-Parmer™ Polypropylene 3-Way Two-Position Elliptic Valve
570.00 DKK
Specifications
Product Type | 3-Way Elliptic Valve |
---|---|
Material | Polypropylene |
For Use With (Application) | For liquid, gas, and vacuum applications |
Packaging Quantity | 1 Pack |
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
11838073
|
Cole-Parmer™
98150-00 |
For 1/8 in. NPT(F) connectors |
570.00 DKK
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
11848073
|
Cole-Parmer™
98150-01 |
For 1/4 in. NPT(F) connectors |
570.00 DKK
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Specifications
3-Way Elliptic Valve | |
Polypropylene | |
For liquid, gas, and vacuum applications | |
1 Pack |