Learn More
Macherey-Nagel™ Polygram CEL 400-10 Glass Plates
Brand:  Macherey-Nagel™ 808073
 View more versions of this product
Product Code. 10236191
  • 1460.00 DKK / Pack of 25
Only null left
Estimated Shipment: 17-07-2025
Specifications
25 Pk.
Glass Plate
0.1 mm
20 x 20 cm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Macherey-Nagel™ Polygram CEL 400-10 Glass Plates > Plate Size: 200 x 200mm