missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygoprep™ 60 to 80

Brand:  Macherey-Nagel™ 711270.5000

 View more versions of this product

Product Code. 10135171

  • 6045.00 DKK / 5kg
Only null left
Estimated Shipment: 27-06-2025
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

63 to 100 ÎĽm
POLYGOPREP 60 to 80
2 to 8
POLYGOPREP
Polygoprep
Liquid Chromatography
60 Ă…
Liquid Chromatography
5 kg
350 m2/g
Silica
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ Polygoprep™ 60 to 80 > Quantity: 5kg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.