Få mere at vide
Pal International™ Deluxe Smooth Sole Fabric Overshoes
Farve:
White
Pakningsstørrelse:
Pakke med 200
Pakke med 250
Beskrivelse
- Manufactured from strong and durable non-woven polypropylene.
- Higher spec overshoe designed for a better fit and improved comfort.
- Especially suitable for more sensitive areas such as labarotories or for visitors to hygienic manufacturing sites.
- Embossed sole gives extra grip and durability.
- Elasticized band for easy wear and removal
- DIsposable
- One size fits all.
- Package of 200
Tekniske data
Tekniske data
| Farve | White |
| Hæl stil | Elasticated ankle |
| Materiale | Polypropylene |
| Størrelse | Universal |
| Sål funktioner | Smooth |
| Produkttype | Overshoe |