missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol methyl ether acrylate, M.W. 1,000, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
Beschreibung
Polyethylene glycol methyl ether acrylate is used as a raw material in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Color | White |
Physical Form | Granular Solid |
Solubility Information | Soluble in water |
Quantity | 250 mg |
Chemical Name or Material | Polyethylene glycol methyl ether acrylate, M.W. 1,000 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'