Få mere at vide
Polyethylene Barrel Tap
Beskrivelse
Two popular sized drum taps with efficient closing for safety purposes. They are particularly appropriate where containers are stored horizontally. Constructed in high density polyethylene they are resistant to a wide range of chemicals. The materials used meet FDA requirements. The 2 inch size is particularly useful for viscous liquids and for 205L drums, whereas the 3/4 inch tap fits smaller containers. In the case of the 2 inch tap, an adapter is offered which converts the BSP thread to Tri-sure for use with containers where this thread is fitted.
Tekniske data
Tekniske data
| Oversigt | 3/4 tommer (1,905 cm) |
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.