Få mere at vide
Brady™ Polyester termotransfer-printbare etiketter
Artikelnummer. 12414890
Skift visning
Form:
Rektangel
Rektangel med afrundede hjørner
Rund
Størrelse (metrisk):
101,6 B x 50,8 mm H
15,24 B x 38,1 mm H
15,24 mm dia.
15,24 x 48,26 mm (B x H)
15,24 x 66,68 mm (B x H)
18,26 B x 25,46 mm H
19 x 6,35 mm (B x H)
19B x 38,1 mmH
19B x 6,35 mmH
22,86 B x 19 mm H
22,86 B x 22,86 mm H
25,4 B x 12,7 mm H
25,4 B x 4,75 mm H
25,4 B x 76,2 mm H
25,4 B x 9,53 mm H
25,4 x 12,70 mm (B x H)
31,75 B x 19 mm H
34,93 B x 45,72 mm H
38,1 B x 19 mm H
38,10 B x 6,35 mm H
40 W x 15 mmH
41,28 x 15,24 mm (B x H)
48,26 B x 6,35 mm H
5,08 B x 22,86 mm H
50,8 B x 25,4 mm H
50,8 B x 6,35 mm H
50,8 x 25,4 mm (B x H)
57,15 B x 19 mm H
76,2 B x 25,4 mm H
76,20 B x 25,4 mm H
76,20 B x 50,8 mm H
88,9 B x 38,1 mm H
9,53 mm dia.
Slutte:
Glans
Mat
Satin
Klæbende type:
Permanent akryl, Permanent
Unit Size:
1 pakke
1 rulning
Hver
34 product options available for selection
Product selection table with 34 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Form Størrelse (metrisk) Slutte Klæbende type unitSize
12414890 Rektangel 34,93 B x 45,72 mm H Mat Permanent akryl, Permanent Hver
13099262 Rektangel 101,6 B x 50,8 mm H Glans Permanent akryl, Permanent 1 rulning
10479883 Rund 9,53 mm dia. Mat Permanent akryl, Permanent 1 rulning
11504314 Rektangel med afrundede hjørner 40 W x 15 mmH Mat Permanent akryl, Permanent 1 rulning
13019302 Rektangel 50,8 B x 6,35 mm H Glans Permanent akryl, Permanent 1 rulning
11896023 Rektangel 19B x 38,1 mmH Mat Permanent akryl, Permanent 1 rulning
13099302 Rektangel 38,10 B x 6,35 mm H Glans Permanent akryl, Permanent 1 rulning
11810855 Rektangel 19 x 6,35 mm (B x H) Glans Permanent akryl, Permanent 1 rulning
11816043 Rektangel 22,86 B x 22,86 mm H Mat Permanent akryl, Permanent 1 rulning
11583784 Rektangel 25,4 B x 4,75 mm H Glans Permanent akryl, Permanent -
10209922 Rektangel med afrundede hjørner 25,4 B x 9,53 mm H Mat Permanent akryl, Permanent 1 rulning
11836013 Rektangel 5,08 B x 22,86 mm H Mat Permanent akryl, Permanent 1 rulning
13009312 Rektangel 25,4 B x 12,7 mm H Glans Permanent akryl, Permanent 1 pakke
12834723 Rektangel 50,8 B x 25,4 mm H Satin Permanent akryl, Permanent 1 rulning
10020942 Rektangel 15,24 x 48,26 mm (B x H) Mat Permanent akryl, Permanent 1 rulning
11538754 Rektangel 25,4 B x 76,2 mm H Satin Permanent akryl, Permanent 1 rulning
11836023 Rektangel 34,93 B x 45,72 mm H Mat Permanent akryl, Permanent 1 rulning
11523724 Rektangel 76,20 B x 25,4 mm H Glans Permanent akryl, Permanent 1 rulning
10011932 Rektangel med afrundede hjørner 15,24 mm dia. Mat Permanent akryl, Permanent 1 rulning
11814714 Rektangel 76,20 B x 50,8 mm H Glans Permanent akryl, Permanent 1 rulning
11503734 Rektangel 31,75 B x 19 mm H Glans Permanent akryl, Permanent 1 rulning
11836043 Rektangel 38,1 B x 19 mm H Satin Permanent akryl, Permanent 1 rulning
10031932 Rektangel 15,24 B x 38,1 mm H Mat Permanent akryl, Permanent 1 rulning
10630134 Rektangel 50,8 x 25,4 mm (B x H) Glans Permanent akryl, Permanent 1 rulning
10315992 Rektangel 48,26 B x 6,35 mm H Mat Permanent akryl, Permanent 1 rulning
11885245 Rektangel 25,4 x 12,70 mm (B x H) Glans Permanent akryl, Permanent 1 rulning
10600784 Rektangel 41,28 x 15,24 mm (B x H) Mat Permanent akryl, Permanent 1 rulning
11582715 Rektangel 76,2 B x 25,4 mm H Glans Permanent akryl, Permanent -
10679343 Rektangel 88,9 B x 38,1 mm H Mat Permanent akryl, Permanent 1 rulning
10561515 Rektangel 18,26 B x 25,46 mm H Mat Permanent akryl, Permanent 1 rulning
11886013 Rektangel 15,24 x 66,68 mm (B x H) Mat Permanent akryl, Permanent 1 rulning
12882699 Rektangel 22,86 B x 19 mm H Mat Permanent akryl, Permanent 1 rulning
11826033 Rektangel 57,15 B x 19 mm H Mat Permanent akryl, Permanent 1 rulning
11826043 Rektangel 19B x 6,35 mmH Satin Permanent akryl, Permanent 1 rulning
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 34 options available.
34 options
Product Code. 12414890 Supplier Brady™ Supplier No. KEA70V49NA
Polyester Thermal-Transfer Printable Labels er lavet af permanent polyester (B-423), som er ideel til elektronisk PCB og komponentidentifikation, stregkodeetiketter, mærkeplader og solpanelidentifikation.
  • Blank hvid finish med permanent akryl kan printes med termisk transfer
  • RoHS i overensstemmelse med 2005/618/EF MCV-ændring af RoHS-direktiv 2002/95/EF; UL godkendte; CSA accepteret
  • Opfylder kravene til et halogenfrit materiale i henhold til DIN VDE 0472 del 815
  • Servicetemperatur på -94 til 230 °F (-70 til 110 °C)
  • Kompatible printere: BBP11, BBP12, THT-printer, 25 mm kerne
Specifications
Autoklaverbar Autoklaverbar
Certificeringer/compliance 2005/618/EC MCV-ændring til RoHS-direktivet 2002/95/EC
Oversigt Thermal Transfer Printable Etiketter
Klæbende type Permanent akryl, Permanent
Belægning Ingen
Farve Hvid, Klarhed
Slutte Mat
Format Rul
Materiale Polyester
Printbare sider Enkeltsidet printbar
Print teknologi Termisk overførsel
Produkttype Termisk overførselsprinteretiket
Sterilitet Ikke-steril
Til brug med (applikation) Laboratorieidentifikation såsom hætteglas, centrifugerør, reagensglas, sugerør og objektglas
Til brug med (udstyr) BBP11, BBP12, THT 76 mm kerneprintere
Etiketter pr. række 1
Min. Driftstemperatur (metrisk) -196 °C
Model- BPT-632-461
nr. pr. pakke 2000/Pk.
Mængde 2000
Form Rektangel
Størrelse 34.93 mm (W) x 45.72 mm (H)
Størrelse (metrisk) 34,93 B x 45,72 mm H
Temperaturområde (engelsk) -321 °F til +230 °F
Temperaturområde (metrisk) -196° C til 110° C
Tykkelse (engelsk) 0,0019 in.
Tykkelse (metrisk) 0.012 mm
Lodret gentagelse (metrisk) 41.72 mm
Dimensioner (L x B) 45.72 x 34.93 mm
Længde (engelsk) 1,8 tommer
Bredde (engelsk) 1,38 tommer
Bredde (metrisk) 34.93 mm
Bredde (engelsk) Udskrivbart område 34.93 mm
Bredde (metrisk) Printbart område 34.93 mm
Højde (engelsk) 1,8 tommer
Højde (engelsk) Printbart område 1,57 tommer
Højde (metrisk) 6.35 mm
Højde (metrisk) printbart område 40 mm
Alternativt varenummer BPT-632-461