Learn More
Macherey-Nagel™ Polyester with Glass Fiber prefilter Syringe Filter
Brand: Macherey-Nagel™ 729033.400
Additional Details : Weight : 0.10000kg
Specifications
Black Top, Orange Bottom | |
1/0.45 μm | |
Hydrophilic | |
Syringe Filter |
Polyester with Glass Fiber Prefilter | |
Polar as well as nonpolar solvents | |
400Pack | |
25 mm |