Learn More
Macherey-Nagel™ Polyester with Glass Fiber prefilter Syringe Filter
Brand:  Macherey-Nagel™ 729033.400
Additional Details : Weight : 0.10000kg
Product Code. 11345790
  • 4885.00 DKK / Pack of 400
Estimated Shipment: 15-05-2024
to see stock.
Specifications
Black Top, Orange Bottom
1/0.45 μm
Hydrophilic
Syringe Filter
Polyester with Glass Fiber Prefilter
Polar as well as nonpolar solvents
400Pack
25 mm