Få mere at vide
Masterflex™ Polycarbonate Smart Site Luer Fitting Connection
Trådstørrelse:
Hun x Han Luer Lock
Pakningsstørrelse:
Pakke med 10
Beskrivelse
- Each assembly comes packaged individually and can be sterilized
- Complete your luer fittings systems with these premium connectors
- Included are some special configurations available only in polycarbonate
- All female ports may be used with either slip or luer lock connections
- Plugs and caps are also available
Tekniske data
Tekniske data
| Materiale | Polycarbonate |
| Emballagemængde | 10 Each |
| Trådstørrelse | Hun x Han Luer Lock |
| Produkttype | Smart Site Luer Fitting Connection |